Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TSN Full-length ORF (NP_004613.1, 1 a.a. - 228 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965960 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-960 10 ug
89-965-961 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-960 Supplier Abnova™ Supplier No. H00007247P02S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a DNA-binding protein which specifically recognizes conserved target sequences at the breakpoint junction of chromosomal translocations. Translin polypeptides form a multimeric structure that is responsible for its DNA-binding activity. Recombination-associated motifs and translin-binding sites are present at recombination hotspots and may serve as indicators of breakpoints in genes which are fused by translocations. These binding activities may play a crucial role in chromosomal translocation in lymphoid neoplasms. [provided by RefSeq]

Sequence: MSVSEIFVELQGFLAAEQDIREEIRKVVQSLEQTAREILTLLQGVHQGAGFQDIPKRCLKAREHFGTVKTHLTSLKTKFPAEQYYRFHEHWRFVLQRLVFLAAFVVYLETETLVTREAVTEILGIEPDREKGFHLDVEDYLSGVLILASELSRLSVNSVTAGDYSRPLHISTFINELDSGFRLLNLKNDSLRKRYDGLKYDVKKVEEVVYDLSIRGFNKETAAACVEK

Specifications

Accession Number NP_004613.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7247
Molecular Weight (g/mol) 52.6kDa
Name TSN (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MSVSEIFVELQGFLAAEQDIREEIRKVVQSLEQTAREILTLLQGVHQGAGFQDIPKRCLKAREHFGTVKTHLTSLKTKFPAEQYYRFHEHWRFVLQRLVFLAAFVVYLETETLVTREAVTEILGIEPDREKGFHLDVEDYLSGVLILASELSRLSVNSVTAGDYSRPLHISTFINELDSGFRLLNLKNDSLRKRYDGLKYDVKKVEEVVYDLSIRGFNKETAAACVEK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BCLF-1/REHF-1/TRSLN
Common Name TSN
Gene Symbol TSN
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.