Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TSPAN7 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7083323
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9179SP 25 μg
MAB9179100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9179SP Supplier R&D Systems Supplier No. MAB9179SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TSPAN7/TM4SF2 Monoclonal specifically detects TSPAN7/TM4SF2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen TSPAN7/TM4SF2
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 482618
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P41732
Gene Alias A15, A15CCG-B7, CCG-B7, CD231, CD231 antigen, Cell surface glycoprotein A15, DXS1692E, Membrane component chromosome X surface marker 1, mental retardation, X-linked 58, MRX58, MXS1, MXS1T-cell acute lymphoblastic leukemia associated antigen 1, TALLA-1, T-cell acute lymphoblastic leukemia-associated antigen 1, tetraspanin 7, tetraspanin-7, TM4SF2, TM4SF2b, TM4SF2tetraspanin protein, transmembrane 4 superfamily 2b, Transmembrane 4 superfamily member 2X chromosome, surface marker 1, transmembrane protein A15, Tspan-7
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human TSPAN7 Accession # P41732
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7102
Test Specificity Stains human TSPAN7 transfectants but not irrelevant transfectants in flow cytometry.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.