Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TSPAN8 Full-length ORF (AAH05246.1, 1 a.a. - 237 a.a.) Recombinant Protein MW: 52.5kDa with GST-tag at N-terminal

Catalog No. 89965988 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-988 10 ug
89-965-989 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-988 Supplier Abnova™ Supplier No. H00007103P02S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins. This gene is expressed in different carcinomas. The use of alternate polyadenylation sites has been found for this gene. [provided by RefSeq]

Sequence: MAGVSACIKYSMFTFNFLFWLCGILILALAIWVRISNDSQAIFGSEDVGSSSYVAVDILIAVGAIIMILGFLACCGAIKESRCMLLLFFIGLLLILLLQVATGILGAVFKSKSDRIVNETLYENTKLLSATGESEKQFQEAIIVFQEEFKCCGLVNGAADWGNNFQHYPELCACLDKQRPCQSYNGKQVYKETCISFIKDFLAKNLIIVIGIAFGLAVIEILGLVFSMVLYCQIGNK

Specifications

Accession Number AAH05246.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7103
Molecular Weight (g/mol) 52.5kDa
Name TSPAN8 (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MAGVSACIKYSMFTFNFLFWLCGILILALAIWVRISNDSQAIFGSEDVGSSSYVAVDILIAVGAIIMILGFLACCGAIKESRCMLLLFFIGLLLILLLQVATGILGAVFKSKSDRIVNETLYENTKLLSATGESEKQFQEAIIVFQEEFKCCGLVNGAADWGNNFQHYPELCACLDKQRPCQSYNGKQVYKETCISFIKDFLAKNLIIVIGIAFGLAVIEILGLVFSMVLYCQIGNK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CO-029/TM4SF3
Common Name TSPAN8
Gene Symbol TSPAN8
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.