Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TSTA3 Partial ORF (NP_003304, 222 a.a. - 321 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966033 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-033 25 ug
89-966-032 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-966-033 Supplier Abnova™ Supplier No. H00007264Q01L

Used for AP, Array, ELISA, WB-Re

Tissue specific transplantation antigen P35B is a NADP(H)-binding protein. It catalyze the two-step epimerase and the reductase reactions in GDP-D-mannose metabolism, converting GDP-4-keto-6-D-deoxymannose to GDP-L-fucose. GDP-L-fucose is the substrate of several fucosyltransferases involved in the expression of many glycoconjugates, including blood group ABH antigens and developmental adhesion antigens. Mutations in this gene may cause leukocyte adhesion deficiency, type II. [provided by RefSeq]

Sequence: DLAQLFIWVLREYNEVEPIILSVGEEDEVSIKEAAEAVVEAMDFHGEVTFDTTKSDGQFKKTASNSKLRTYLPDFRFTPFKQAVKETCAWFTDNYEQARK

Specifications

Accession Number NP_003304
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7264
Molecular Weight (g/mol) 36.74kDa
Name TSTA3 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen DLAQLFIWVLREYNEVEPIILSVGEEDEVSIKEAAEAVVEAMDFHGEVTFDTTKSDGQFKKTASNSKLRTYLPDFRFTPFKQAVKETCAWFTDNYEQARK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FX/P35B/SDR4E1
Common Name TSTA3
Gene Symbol TSTA3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.