Learn More
Invitrogen™ Human Tsukushi/TSK ELISA Kit
Description
Human Tsukushi/TSK quantitates human Tsukushi/TSK in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Tsukushi/TSK.
Specifications
Specifications
| Accession Number | 9530051K01Rik; E2IG4; E2-induced gene 4 protein; Early insulin-induced hepatic gene protein; EIIH; hepatic protein EIIH; leucine rich repeat containing 54; leucine-rich repeat-containing protein 54; Lrrc54; Tsk; Tsku; tsukushi; tsukushi homolog; tsukushi small leucine rich proteoglycan homolog; tsukushi, small leucine rich proteoglycan; Tsukushin; UNQ850/PRO1788 |
| Assay Range | 0.63 to 150 ng/mL |
| Assay Sensitivity | 0.63 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 25987 |
| Gene Symbol | TSKU |
| Interassay CV | <12% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.