Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Tsukushi/TSK ELISA Kit

Catalog No. EH474RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH474RBX10 10 x 96 Tests
EH474RBX5 5 x 96 Tests
EH474RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH474RBX10 Supplier Invitrogen™ Supplier No. EH474RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Human Tsukushi/TSK quantitates human Tsukushi/TSK in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Tsukushi/TSK.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 9530051K01Rik; E2IG4; E2-induced gene 4 protein; Early insulin-induced hepatic gene protein; EIIH; hepatic protein EIIH; leucine rich repeat containing 54; leucine-rich repeat-containing protein 54; Lrrc54; Tsk; Tsku; tsukushi; tsukushi homolog; tsukushi small leucine rich proteoglycan homolog; tsukushi, small leucine rich proteoglycan; Tsukushin; UNQ850/PRO1788
Assay Range 0.63 to 150 ng/mL
Assay Sensitivity 0.63 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 25987
Gene Symbol TSKU
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target TSKU
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.