Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human TWEAK, Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF3100625UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3100625UG 25 μg
AF31006100 100 μg
AF310061MG 1 mg
AF31006500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AF3100625UG Supplier Gibco™ Supplier No. AF3100625UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human TWEAK is a soluble 17.0 kDa polypeptide (154 amino acid residues) comprising the TNF-homologous region of TWEAK, and is generated by proteolytic processing of the full length membrane-anchored TWEAK protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TNF-related weak inducer of apoptosis (TWEAK) is a member of the tumor necrosis factor superfamily (TNFSF) of structurally related cytokines. Like most other members of this family, TWEAK is a cell surface-associated type II transmembrane protein although a smaller, biologically active form can also be generated by cleavage near the cell membrane. TWEAK has multiple biological activities, including stimulation of cell growth and angiogenesis, induction of inflammatory cytokines, in addition to stimulation of apoptosis. The TWEAK signal transduction pathway has not been well established but it appears to signal via TweakR/Fn14 in a manner similar to that described for other TNFSF members that bind receptors lacking death domains.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43508
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 8742
Molecular Weight (g/mol) 17 kDa
Name Human TWEAK, Animal-Free
Quantity 25 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias APO3 ligand; APO3/DR3 ligand; APO3L; Dr3l; Dr3lg; LOW QUALITY PROTEIN: tumor necrosis factor ligand superfamily member 12; MGC129581; MGC20669; TNF superfamily member 12; TNF-related weak inducer of apoptosis; TNFSF12; TNLG4A; tumor necrosis factor (ligand) superfamily member 12; tumor necrosis factor (ligand) superfamily, member 12; tumor necrosis factor ligand 4 A; tumor necrosis factor ligand superfamily member 12; Tumor necrosis factor ligand superfamily member 12, membrane form; Tumor necrosis factor ligand superfamily member 12, secreted form; tumor necrosis factor superfamily member 12; TWEAK; UNQ181/PRO207; zgc:153941
Common Name TWEAK
Gene Symbol Tnfsf12
Biological Activity Assay 1: The ED50 as determined by the dose-dependent stimulation of IL-8 production by human PBMC is less than 10 ng/ml Assay 2: TWEAK weakly induces the death of HT29 cells when cultured in the presence of IFN-gamma. The ED50 for this effect is between 30-45 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence MKGRKTRARR AIAAHYEVHP RPGQDGAQAG VDGTVSGWEE ARINSSSPLR YNRQIGEFIV TRAGLYYLYC QVHFDEGKAV YLKLDLLVDG VLALRCLEEF SATAASSLGP QLRLCQVSGL LALRPGSSLR IRTLPWAHLK AAPFLTYFGL FQVH
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.