Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TXK (NM_003328, 239 a.a. - end) Partial Recombinant Protein with GST-tag expressed in Sf9 cells

Catalog No. 89966238 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-238 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-966-238 Supplier Abnova™ Supplier No. P3927

Used for Func, SDS-PAGE

Specifications

Accession Number NM_003328
For Use With (Application) Functional Study, SDS-PAGE
Formulation In 50mM Tris-HCl, 150mM NaCl, pH 7.5 (0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol)
Gene ID (Entrez) 7294
Molecular Weight (g/mol) 53kDa
Name TXK (Human) Recombinant Protein
Preparation Method Insect cell (Sf9) expression system
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BTKL/MGC22473/PSCTK5/PTK4/RLK/TKL
Common Name TXK
Gene Symbol TXK
Biological Activity The specific activity was determined to be 18nmol /min/mg as per Activity Assay Protocol.
Species Baculovirus
Recombinant Recombinant
Protein Tag GST
Expression System Insect cell (Sf9) expression system
Purity or Quality Grade >90% by densitometry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.