Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human UbcH5a/UBE2D1 - Ubiquitin Charged Protein

Catalog No. E2800100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E2800100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E2800100 Supplier R&D Systems™ Supplier No. E2800100
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Pre-charged UBE2D1 may be added directly to in vitro reactions containing E3 Ubiquitin ligases and ligase substrates (if applicable)—no ATP, E1 enzyme, or extra Ubiquitin are required. Reaction conditions will need to be optimized for each specific application. Note: Reducing agents including dithiothreitol (DTT) or mercaptoethanol ( beta ME) may cause unintended thiolytic release of Ubiquitin from the complex—care must be taken if these compounds are present in buffers. We suggest using a thioester friendly reductant such as TCEP if possible.

Specifications

Accession Number P51668, P0CG47
For Use With (Application) Bioactivity
Formulation Approximately 0.43 mg/mL (25 μM) with respect to UBE2D1. Formulated in 50 mM HEPES pH 7.5, 50 mM NaCl, 1 mM TCEP
Gene ID (Entrez) 7321
Molecular Weight (g/mol) M.W. (theoretical): 25 kDa (17 kDa UBE2D1, 8.6 kDa Ubiquitin)
Quantity 100 μg
Source E. coli-derived human UbcH5a/UBE2D1 protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Alias E2(17)KB1, Stimulator of Fe transportUBC4/5 homolog, UBC4/5, UBC5A, UbcH5, UbcH5A, UBCH5AEC 6.3.2.19, UBCH5SFT, UBE2D1, Ubiquitin carrier protein D1, ubiquitin-conjugating enzyme E2 D1, Ubiquitin-conjugating enzyme E2(17)KB 1, Ubiquitin-conjugating enzyme E2-17 kDa 1, ubiquitin-conjugating enzyme E2D 1 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase D1
Common Name UbcH5a/UBE2D1
Gene Symbol UBE2D1
Biological Activity Pre-charged UBE2D1 may be added directly to in vitro reactions containing E3 Ubiquitin ligases and ligase substrates (if applicable)—no ATP, E1 enzyme, or extra Ubiquitin are required. Reaction conditions will need to be optimized for each specific application. Note: Reducing agents including dithiothreitol (DTT) or mercaptoethanol ( beta ME) may cause unintended thiolytic release of Ubiquitin from the complex—care must be taken if these compounds are present in buffers. We suggest using a thioester friendly reductant such as TCEP if possible.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Accession # P51668, P0CG47
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein UbcH5a/UBE2D1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.