Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UBE3B Partial ORF (NP_542783, 118 a.a. - 225 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966439 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-439 10 μg
89-966-440 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-966-439 Supplier Abnova™ Supplier No. H00089910Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E3 ubiquitin-conjugating enzyme family. The encoded protein may interact with other proteins and play a role in stress response. Alternatively spliced transcript variants encoding the same protein isoform have been identified for this gene. [provided by RefSeq]

Sequence: VSLACSKDLTLLWIQQIKNILWYCCDFLKQLKPEILQDSRLITLYLTMLVTFTDTSTWKILRGKGESLRPAMNHICANIMGHLNQHGFYSVLQILLTRGLARPRPCLS

Specifications

Accession Number NP_542783
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 89910
Molecular Weight (g/mol) 37.62kDa
Name UBE3B (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen VSLACSKDLTLLWIQQIKNILWYCCDFLKQLKPEILQDSRLITLYLTMLVTFTDTSTWKILRGKGESLRPAMNHICANIMGHLNQHGFYSVLQILLTRGLARPRPCLS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp586K2123/DKFZp686A1051/FLJ45294/MGC131858/MGC78388
Common Name UBE3B
Gene Symbol UBE3B
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.