Learn More
Biomatik Corporation Human Ubiquitin Cross Reactive Protein (UCRP) CLIA Kit
Supplier:  Biomatik Corporation EKU09715
Encompass_Preferred
Human Ubiquitin Cross Reactive Protein (UCRP) CLIA Kit for detecting UCRP in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 11.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P05161. Gene ID: 9636. Target Synonyms: G1P2; IFI15; ISG15; Interferon Alpha-Inducible Protein clone IFI-15K; Interferon Induced Protein 15; Interferon-induced 17 kDa protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4353
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.