Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Ubiquitin Mutant S65A, Carrier-Free Recombinant Protein

Catalog No. UMS65A100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
UMS65A100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. UMS65A100 Supplier R&D Systems™ Supplier No. UMS65A100
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Ubiquitin Mutant S65A Protein is derived from E. coli. The Recombinant Human Ubiquitin Mutant S65A Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P0CG47
For Use With (Application) Bioactivity
Gene ID (Entrez) 6233
Molecular Weight (g/mol) Theoretical M.W.: 8.6kDa
Quantity 100 μg
Source E. coli-derived human Ubiquitin protein Contains a Ser-to-Ala substitution at position 65.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias RPS27A, UBA52, UBB, UBB ubiquitin B, UBC
Common Name Ubiquitin
Gene Symbol UBB
Biological Activity Recombinant Ubiquitin Mutant S65A may be used as a negative control in experiments examining PINK1 kinase activity. Ubiquitin S65A is not phosphorylated by PINK1 in vitro. Reaction conditions will need to be optimized for each specific application.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Contains a Ser-to-Ala substitution at position 65.
Purity or Quality Grade >98%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.