Learn More
Biomatik Corporation Human Ubiquitin Specific Peptidase 11 (USP11) ELISA Kit
Supplier:  Biomatik Corporation EKU12077
Encompass_Preferred
Human Ubiquitin Specific Peptidase 11 (USP11) ELISA Kit for detecting USP11 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51784. Target Synonyms: UHX1; Ubiquitin carboxyl-terminal hydrolase 11; Deubiquitinating enzyme 11; Ubiquitin thioesterase 11; Ubiquitin-specific-processing protease 11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8039
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.