Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UBR1 Partial ORF (NP_777576, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966471 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-471 25 μg
89-966-470 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-966-471 Supplier Abnova™ Supplier No. H00197131Q01L

Used for AP, Array, ELISA, WB-Re

The N-end rule pathway is one proteolytic pathway of the ubiquitin system. The recognition component of this pathway, encoded by this gene, binds to a destabilizing N-terminal residue of a substrate protein and participates in the formation of a substrate-linked multiubiquitin chain. This leads to the eventual degradation of the substrate protein. The protein described in this record has a RING-type zinc finger and a UBR-type zinc finger. Mutations in this gene have been associated with Johanson-Blizzard syndrome. [provided by RefSeq]

Sequence: ADEEAGGTERMEISAELPQTPQRLASWWDQQVDFYTAFLHHLAQLVPEIYFAEMDPDLEKQEESVQMSIFTPLEWYLFGEDPDICLEKLKHSGAFQLCG

Specifications

Accession Number NP_777576
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 197131
Molecular Weight (g/mol) 36.63kDa
Name UBR1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen ADEEAGGTERMEISAELPQTPQRLASWWDQQVDFYTAFLHHLAQLVPEIYFAEMDPDLEKQEESVQMSIFTPLEWYLFGEDPDICLEKLKHSGAFQLCG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias JBS/MGC142065/MGC142067
Common Name UBR1
Gene Symbol UBR1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.