Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UBR2 Full-length ORF (AAH64512.1, 1 a.a. - 439 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966474 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-474 10 μg
89-966-475 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-966-474 Supplier Abnova™ Supplier No. H00023304P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Proteolysis by the ubiquitin-proteasome system controls the concentration of many regulatory proteins. The selectivity of ubiquitylation is determined by ubiquitin E3 ligases, which recognize the substrate's destabilization signal, or degron. The E3 ligase UBR2 participates in the N-end rule pathway, which targets proteins bearing an N-terminal degron, or N-degron (Kwon et al., 2003 [PubMed 14585983]).[supplied by OMIM]

Sequence: MASELEPEVQAIDRSLLECSAEEIAGKWLQATDLTREVYQHLAHYVPKIYCRGPNPFPQKEDMLAQHVLLGPMEWYLCGEDPAFGFPKLEQANKPSHLCGRVFKVGEPTYSCRDCAVDPTCVLCMECFLGSIHRDHRYRMTTSGGGGFCDCGDTEAWKEGPYCQKHELNTSEIEEEEDPLVHLSEDVIARTYNIFAITFRYAVEILTWEKESELPADLEMVEKSDTYYCMLFNDEVHTYEQVIYTLQKAVNCTQKEAIGFATTVDRDGRRSVRYGDFQYCEQAKSVIVRNTSRQTKPLKVQVMHSSIVAHQNFGLKLLSWLGSIIGYSDGLRRILCQVGLQEGPDGENSSLVDRLMLSDSKLWKGARSVYHQLFMSSLLMDLKYKKLFAVRFAKNYERLQSDYVTDDHDREFSVADLSVQIFTVPSLFSISAGRSGSPL

Specifications

Accession Number AAH64512.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23304
Molecular Weight (g/mol) 76.4kDa
Name UBR2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MASELEPEVQAIDRSLLECSAEEIAGKWLQATDLTREVYQHLAHYVPKIYCRGPNPFPQKEDMLAQHVLLGPMEWYLCGEDPAFGFPKLEQANKPSHLCGRVFKVGEPTYSCRDCAVDPTCVLCMECFLGSIHRDHRYRMTTSGGGGFCDCGDTEAWKEGPYCQKHELNTSEIEEEEDPLVHLSEDVIARTYNIFAITFRYAVEILTWEKESELPADLEMVEKSDTYYCMLFNDEVHTYEQVIYTLQKAVNCTQKEAIGFATTVDRDGRRSVRYGDFQYCEQAKSVIVRNTSRQTKPLKVQVMHSSIVAHQNFGLKLLSWLGSIIGYSDGLRRILCQVGLQEGPDGENSSLVDRLMLSDSKLWKGARSVYHQLFMSSLLMDLKYKKLFAVRFAKNYERLQSDYVTDDHDREFSVADLSVQIFTVPSLFSISAGRSGSPL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias C6orf133/DKFZp686C08114/KIAA0349/MGC71112/RP3-392M17.3/bA49A4.1/dJ242G1.1/dJ392M17.3
Common Name UBR2
Gene Symbol UBR2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.