Learn More
Abnova™ Human UBR2 Partial ORF (NP_056070, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
Proteolysis by the ubiquitin-proteasome system controls the concentration of many regulatory proteins. The selectivity of ubiquitylation is determined by ubiquitin E3 ligases, which recognize the substrate's destabilization signal, or degron. The E3 ligase UBR2 participates in the N-end rule pathway, which targets proteins bearing an N-terminal degron, or N-degron (Kwon et al., 2003 [PubMed 14585983]).[supplied by OMIM]
Specifications
Specifications
| Accession Number | NP_056070 |
| For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 23304 |
| Molecular Weight (g/mol) | 36.74kDa |
| Name | UBR2 (Human) Recombinant Protein (Q01) |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 25 μg |
| Immunogen | MASELEPEVQAIDRSLLECSAEEIAGKWLQATDLTREVYQHLAHYVPKIYCRGPNPFPQKEDMLAQHVLLGPMEWYLCGEDPAFGFPKLEQANKPSHLCG |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.