Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UBXD5 Partial ORF (NP_663320.1, 311 a.a. - 409 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89962802 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-802 10 μg
89-962-803 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-802 Supplier Abnova™ Supplier No. H00091544Q01S

Used for AP, Array, ELISA, WB-Re

This gene encodes a protein with a divergent C-terminal UBX domain. The homologous protein in the rat interacts with members of the Rnd subfamily of Rho GTPases at the cell periphery through its C-terminal region. It also interacts with several heterotrimeric G proteins through their G-alpha subunits and promotes Rho GTPase activation. It is proposed to serve a bidirectional role in the promotion and inhibition of Rho activity through upstream signaling pathways. The 3' coding sequence of this gene contains a polymoprhic region of 24 nt tandem repeats. Several transcripts containing between 1.5 and five repeat units have been reported. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: QGEVIDIRGPIRDTLQNCCPLPARIQEIVVETPTLAAERERSQESPNTPAPPLSMLRIKSENGEQAFLLMMQPDNTIGDVRALLAQARVMDASAFEIFS

Specifications

Accession Number NP_663320.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 91544
Molecular Weight (g/mol) 36.41kDa
Name UBXD5 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen QGEVIDIRGPIRDTLQNCCPLPARIQEIVVETPTLAAERERSQESPNTPAPPLSMLRIKSENGEQAFLLMMQPDNTIGDVRALLAQARVMDASAFEIFS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias COA-1/DKFZp686F04228/PP2243/SOC/SOCI/UBXD5
Common Name UBXN11
Gene Symbol UBXN11
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.