Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human UCH-L5/UCH37 Protein, CF

Catalog No. E327025
Change view
Click to view available options
25μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
E327025 25μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E327025 Supplier R&D Systems™ Supplier No. E327025
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Reaction conditions will need to be optimized for each specific application. We recommend an initial recombinant human UCH-L5 concentration of 20-100 nM when using Ubiquitin-AMC or Ubiquitin-Rh110 substrates (U-550, U-555)

Specifications

Accession Number Q9Y5K5
For Use With (Application) Bioactivity
Formulation X mg/mL (X μM) in 50 mM HEPES pH 7.5, 200 mM NaCl, 1 mM TCEP, 10% (v/v) Glycerol
Gene ID (Entrez) 51377
Molecular Weight (g/mol) M.W. (theoretical): 38 kDa
Quantity 25 μg
Source E. coli-derived human UCH-L5/UCH37 protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Alias CGI-70, EC 3.4.19.12, INO80 complex subunit R, INO80R, ubiquitin carboxyl-terminal esterase L5, ubiquitin carboxyl-terminal hydrolase isozyme L5, ubiquitin carboxyl-terminal hydrolase L5, Ubiquitin C-terminal hydrolase UCH37, Ubiquitin thioesterase L5, UCH37, UCH37CGI-70, UCHL5, UCH-L5
Common Name UCH-L5/UCH37
Gene Symbol UCHL5
Biological Activity Reaction conditions will need to be optimized for each specific application. We recommend an initial recombinant human UCH-L5 concentration of 20-100 nM when using Ubiquitin-AMC or Ubiquitin-Rh110 substrates (U-550, U-555)
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein UCH-L5/UCH37
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.