Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UCK1 Partial ORF (NP_113620, 202 a.a. - 276 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966506 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-506 25 μg
89-966-505 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-966-506 Supplier Abnova™ Supplier No. H00083549Q01L

Used for AP, Array, ELISA, WB-Re

Uridine/cytidine kinase-1 (EC 2.7.1.48) is a pyrimidine ribonucleoside kinase that catalyzes the phosphorylation of uridine and cytidine to form uridine monophosphate (UMP) and cytidine monophosphate (CMP) (Van Rompay et al., 2001 [PubMed 11306702]).[supplied by OMIM]

Sequence: TKKYADVIIPRGVDNMVAINLIVQHIQDILNGDICKWHRGGSNGRSYKRTFSEPGDHPGMLTSGKRSHLESSSRP

Specifications

Accession Number NP_113620
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 83549
Molecular Weight (g/mol) 33.99kDa
Name UCK1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen TKKYADVIIPRGVDNMVAINLIVQHIQDILNGDICKWHRGGSNGRSYKRTFSEPGDHPGMLTSGKRSHLESSSRP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ12255/URK1
Common Name UCK1
Gene Symbol UCK1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.