Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UHRF2 Partial ORF (NP_690856, 81 a.a. - 180 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966595 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-595 25 μg
89-966-594 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-966-595 Supplier Abnova™ Supplier No. H00115426Q01L

Used for AP, Array, ELISA, WB-Re

This gene encodes a nuclear protein which is involved in cell-cycle regulation. The encoded protein is a ubiquitin-ligase capable of ubiquinating PCNP (PEST-containing nuclear protein), and together they may play a role in tumorigenesis. [provided by RefSeq]

Sequence: PGTSTQIEAKPCSNSPPKVKKAPRVGPSNQPSTSARARLIDPGFGIYKVNELVDARDVGLGAWFEAHIHSVTRASDGQSRGKTPLKNGSSCKRTNGNIKH

Specifications

Accession Number NP_690856
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 115426
Molecular Weight (g/mol) 36.74kDa
Name UHRF2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen PGTSTQIEAKPCSNSPPKVKKAPRVGPSNQPSTSARARLIDPGFGIYKVNELVDARDVGLGAWFEAHIHSVTRASDGQSRGKTPLKNGSSCKRTNGNIKH
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp434B0920/DKFZp686G0837/MGC33463/NIRF/RNF107/URF2
Common Name UHRF2
Gene Symbol UHRF2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.