Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ULBP-2/5/6 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1298P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1298P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1298P Supplier R&D Systems Supplier No. FAB1298P
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 15 publications

ULBP-2/5/6 Monoclonal specifically detects ULBP-2/5/6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ULBP-2/5/6
Applications Flow Cytometry
Classification Monoclonal
Clone 165903
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9BZM5
Gene Alias ALCAN-alpha, N2DL2, NKG2DL2, RAET1G, RAET1H, RAET1L, UL16 binding protein 2, ULBP2, ULBP5, ULBP6
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human ULBP-2 Accession # Q9BZM5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80328
Test Specificity Detects human ULBP-2 and human RAET1L/ULBP-6 in direct ELISA and stains cells transfected with human ULBP-2, human ULBP-5 or human RAET1L/ULBP-6 in flow cytometry. In direct ELISA, 11% cross-reactivity with recombinant human ULBP5 is observed. It does not stain cells transfected with human ULBP-1 or human ULBP-3 in flow cytometry.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.