Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UNC93B1 Partial ORF (NP_112192, 83 a.a. - 135 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966662 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-662 10 μg
89-966-663 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-966-662 Supplier Abnova™ Supplier No. H00081622Q01S

Used for AP, Array, ELISA, WB-Re

This gene encodes a protein with similarity to the C. elegans unc93 protein. The Unc93 protein is involved in the regulation or coordination of muscle contraction in the worm. [provided by RefSeq]

Sequence: QMQLILHYDETYREVKYGNMGLPDIDSKMLMGINVTPIAALLYTPVLIRFFGT

Specifications

Accession Number NP_112192
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 81622
Molecular Weight (g/mol) 31.57kDa
Name UNC93B1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen QMQLILHYDETYREVKYGNMGLPDIDSKMLMGINVTPIAALLYTPVLIRFFGT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC126617/UNC93/UNC93B
Common Name UNC93B1
Gene Symbol UNC93B1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.