Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UPK3B Full-length ORF (NP_085047.1, 1 a.a. - 320 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966704 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-704 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-966-704 Supplier Abnova™ Supplier No. H00080761P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

UPK3B is a minor component of the apical plaques of mammalian urothelium that binds and dimerizes with uroplakin-1b (UPK1B; MIM 602380), one of the major conserved urothelium membrane proteins. The other major conserved integral membrane proteins of urothelial plaques are UPK1A (MIM 611557), UPK2 (MIM 611558), and UPK3A (MIM 611559) (Deng et al., 2002 [PubMed 12446744]).[supplied by OMIM]

Sequence: MGLPWGQPHLGLQMLLLALNCLRPSLSLGEWGSWMDASSQTQGAGGPAGVIGPWAPAPLRLGEAAPGTPTPVSVAHLLSPVATELVPYTPQITAWDLEGKVTATTFSLEQPRCVFDGLASASDTVWLVVAFSNASRGFQNPETLADIPASPQLLTDGHYMTLPLSPDQLPCGDPMAGSGGAPVLRVGHDHGCHQQPFCNAPLPGPGPYREDPRIHRHLARAAKWQHDRHYLHPLFSGRPPTLGLLGSLYHALLQPVVAGGGPGAAADRLLHGQALHDPPHPTQRGRHTAGGLQAWPGPPPQPQPLAWPLCMGLGEMGRRE

Specifications

Accession Number NP_085047.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 80761
Molecular Weight (g/mol) 60.3kDa
Name UPK3B (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MGLPWGQPHLGLQMLLLALNCLRPSLSLGEWGSWMDASSQTQGAGGPAGVIGPWAPAPLRLGEAAPGTPTPVSVAHLLSPVATELVPYTPQITAWDLEGKVTATTFSLEQPRCVFDGLASASDTVWLVVAFSNASRGFQNPETLADIPASPQLLTDGHYMTLPLSPDQLPCGDPMAGSGGAPVLRVGHDHGCHQQPFCNAPLPGPGPYREDPRIHRHLARAAKWQHDRHYLHPLFSGRPPTLGLLGSLYHALLQPVVAGGGPGAAADRLLHGQALHDPPHPTQRGRHTAGGLQAWPGPPPQPQPLAWPLCMGLGEMGRRE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ32198/MGC10902/P35/UPIIIb
Common Name UPK3B
Gene Symbol UPK3B
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.