Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UPP1 Partial ORF (NP_003355, 1 a.a. - 92 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966705 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-705 10 μg
89-966-706 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-966-705 Supplier Abnova™ Supplier No. H00007378Q01S

Used for AP, Array, ELISA, WB-Re

The 2 known types of pyrimidine nucleoside phosphorylases, uridine phosphorylase (UP; EC 2.4.2.3) and thymidine phosphorylase (TP; EC 2.4.2.4), in the presence of orthophosphate, catalyze the reversible phosphorolysis of uridine and thymidine or deoxyuridine, respectively, to free bases and ribose-1-phosphate or deoxyribose-1-phosphate. Pyrimidine nucleoside phosphorylases can add ribose or deoxyribose to pyrimidine bases to form nucleosides that can be incorporated into RNA or DNA (Watanabe and Uchida, 1995 [PubMed 7488099]).[supplied by OMIM]

Sequence: MAATGANAEKAESHNDCPVRLLNPNIAKMKEDILYHFNLTTSRHNFPALFGDVKFVCVGGSPSRMKAFIRCVGAELGLDCPGRDYPNICAGT

Specifications

Accession Number NP_003355
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7378
Molecular Weight (g/mol) 35.86kDa
Name UPP1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MAATGANAEKAESHNDCPVRLLNPNIAKMKEDILYHFNLTTSRHNFPALFGDVKFVCVGGSPSRMKAFIRCVGAELGLDCPGRDYPNICAGT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias UDRPASE/UP/UPASE/UPP
Common Name UPP1
Gene Symbol UPP1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.