Learn More
R&D Systems™ Human Urinary TIM-1/KIM-1/HAVCR Quantikine ELISA Kit

Description
Specifications
Specifications
| Assay Range | 0.16 to 10 ng/mL |
| Assay Sensitivity | 0.046 ng/mL |
| Conjugate | HRP |
| Product Line | R&D Systems |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Urine |
| Specificity | Natural and Recombinant Human TIM-1 |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 26762 |
| Gene Symbol | HAVCR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.