Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human USH1C Full-length ORF (AAH16057.1, 1 a.a. - 533 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966751 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-751 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-966-751 Supplier Abnova™ Supplier No. H00010083P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a scaffold protein that functions in the assembly of Usher protein complexes. The protein contains PDZ domains, a coiled-coil region with a bipartite nuclear localization signal and a PEST degradation sequence. Defects in this gene are the cause of Usher syndrome type 1C and non-syndromic sensorineural deafness autosomal recessive type 18. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MDRKVAREFRHKVDFLIENDAEKDYLYDVLRMYHQTMDVAVLVGDLKLVINEPSRLPLFDAIRPLIPLKHQVEYDQLTPRRSRKLKEVRLDRLHPEGLGLSVRGGLEFGCGLFISHLIKGGQADSVGLQVGDEIVRINGYSISSCTHEEVINLIRTKKTVSIKVRHIGLIPVKSSPDEPLTWQYVDQFVSESGGVRGSLGSPGNRENKEKKVFISLVGSRGLGCSISSGPIQKPGIFISHVKPGSLSAEVGLEIGDQIVEVNGVDFSNLDHKEGRELFMTDRERLAEARQRELQRQELLMQKRLAMESNKILQEQQEMERQRRKEIAQKAAEENERYRKEMEQIVEEEEKFKKQWEEDWGSKEQLLLPKTITAEVHPVPLRKPKYDQGVEPELEPADDLDGGTEEQGEQDFRKYEEGFDPYSMFTPEQIMGKDVRLLRIKKEGSLDLALEGGVDSPIGKVVVSAVYERGAAERHGGIVKGDEIMAINGKIVTDYTLAEADAALQKAWNQGGDWIDLVVAVCPPKEYDDELTFF

Specifications

Accession Number AAH16057.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10083
Molecular Weight (g/mol) 86.7kDa
Name USH1C (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MDRKVAREFRHKVDFLIENDAEKDYLYDVLRMYHQTMDVAVLVGDLKLVINEPSRLPLFDAIRPLIPLKHQVEYDQLTPRRSRKLKEVRLDRLHPEGLGLSVRGGLEFGCGLFISHLIKGGQADSVGLQVGDEIVRINGYSISSCTHEEVINLIRTKKTVSIKVRHIGLIPVKSSPDEPLTWQYVDQFVSESGGVRGSLGSPGNRENKEKKVFISLVGSRGLGCSISSGPIQKPGIFISHVKPGSLSAEVGLEIGDQIVEVNGVDFSNLDHKEGRELFMTDRERLAEARQRELQRQELLMQKRLAMESNKILQEQQEMERQRRKEIAQKAAEENERYRKEMEQIVEEEEKFKKQWEEDWGSKEQLLLPKTITAEVHPVPLRKPKYDQGVEPELEPADDLDGGTEEQGEQDFRKYEEGFDPYSMFTPEQIMGKDVRLLRIKKEGSLDLALEGGVDSPIGKVVVSAVYERGAAERHGGIVKGDEIMAINGKIVTDYTLAEADAALQKAWNQGGDWIDLVVAVCPPKEYDDELTFF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias AIE-75/DFNB18/NY-CO-37/NY-CO-38/PDZ-45/PDZ-73/PDZ-73/NY-CO-38/PDZ73/ush1cpst
Common Name USH1C
Gene Symbol USH1C
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.