Learn More
Abnova™ Human USO1 Full-length ORF (AAH32654, 1 a.a. - 961 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
The protein encoded by this gene is a peripheral membrane protein which recycles between the cytosol and the Golgi apparatus during interphase. It is regulated by phosphorylation: dephosphorylated protein associates with the Golgi membrane and dissociates from the membrane upon phosphorylation. Ras-associated protein 1 recruits this protein to coat protein complex II (COPII) vesicles during budding from the endoplasmic reticulum, where it interacts with a set of COPII vesicle-associated SNAREs to form a cis-SNARE complex that promotes targeting to the Golgi apparatus. Transport from the ER to the cis/medial Golgi compartments requires the action of this gene product, GM130 and giantin in a sequential manner. [provided by RefSeq]
Specifications
Specifications
| Accession Number | AAH32654 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 8615 |
| Molecular Weight (g/mol) | 131.45kDa |
| Name | USO1 (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue |
| Quantity | 10 μg |
| Immunogen | MNFLRGVMGGQSAGPQHTEAETIQKLCDRVASSTLLDDRRNAVRALKSLSKKYRLEVGIQAMEHLIHVLQTDRSDSEIIGYALDTLYNIISNEEEEVEENSTRQSEDLGSQFTEIFIKQQENVTLLLSLLEEFDFHVRWPGVKLLTSLLKQLGPQVQQIILVSPMGVSRLMDLLADSREVIRNDGVLLLQALTRSNGAIQKIVAFENAFERLLDIISEEGNSDGGIVVEDCLILLQNLLKNNNSNQNFFKEGSYIQRMKPWFEVGDENSGWSAQKVTNLHLMLQLVRVLVSPTNPPGATSSCQKAMFQCGLLQQLCTILMATGVPADILTETINTVSEVIRGCQVNQDYFASVNAPSNPPRPAIVVLLMSMVNERQPFVLRCAVLYCFQCFLYKNQKGQGEIVSTLLPSTIDATGNSVSAGQLLCGGLFSTDSLSNWCAAVALAHALQENATQKEQLLRVQLATSIGNPPVSLLQQCTNILSQGSKIQTRVGLLMLLCTWLSNCPIAVTHFLHNSANVPFLTGQIAENLGEEEQLVQGLCALLLGISIYFNDNSLESYMKEKLKQLIEKRIGKENFIEKLGFISKHELYSRASQKPQPNFPSPEYMIFDHEFTKLVKELEGVITKAIYKSSEEDKKEEEVKKTLEQHDNIVTHYKNMIREQDLQLEELRQQVSTLKCQNEQLQTAVTQQVSQIQQHKDQYNLLKIQLGKDNQHQGSYSEGAQMNGIQPEEIGRLREEIEELKRNQELLQSQLTEKDSMIENMKSSQTSGTNEQSSAIVSARDSEQVAELKQELATLKSQLNSQSVEITKLQTEKQELLQKTEAFAKSVEVQGETETIIATKTTDVEGRLSALLQETKELKNEIKALSEERTAIKEQLDSSNSTIAILQTEKDKLELEITDSKKEQDDLLVLLADQDQKILSLKNKLKDLGHPVEEEDELESGDQEDEDDESEDPGKDLDHI |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.