Learn More
Abnova™ Human USP13 Full-length ORF (CAL38270.1, 1 a.a. - 863 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
Specifications
Specifications
| Accession Number | CAL38270.1 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 8975 |
| Molecular Weight (g/mol) | 121.44kDa |
| Name | USP13 (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 10 μg |
| Immunogen | MQRRGALFGMPGGSGGRKMAAGDIGELLVPHMPTIRVPRSGDRVYKNECAFSYDSPNSEGGLYVCMNTFLAFGREHVERHFRKTGQSVYMHLKRHVREKVRGASGGALPKRRNSKIFLDLDTDDDLNSDDYEYEDEAKLVIFPDHYEIALPNIEELPALVTIACDAVLSSKSPYRKQDPDTWENELPVSKYANNLTQLDNGVRIPPSGWKCARCDLRENLWLNLTDGSVLCGKWFFDSSGGNGHALEHYRDMGYPLAVKLGTITPDGADVYSFQEEEPVLDPHLAKHLAHFGIDMLHMHGPENGLQDNDIKLRVSEWEVIQESGTKLKPMYGPGYTGLKNLGNSCYLSSVMQAIFSIPEFQRAYVGNLPRIFDYSPLDPTQDFNTQMTKLGHGLLSGQYSKPPVKSELIEQVMKEEHKPQQNGISPRMFKAFVSKSHPEFSSNRQQDAQEFFLHLVNLVERNRIGSENPSDVFRFLVEERIQCCQTRKVRYAERVDYLMQLPVAMEAATNKDELIAYELTRREAEANRRPLPELVRAKIPFSACLQAFSEPENVDGFWSSALQAKSAGVKTSRFASFPEYLVVQIKKFTFGLDWVPKKFDVSIDMPDLLDINHLRARGLQPGEEELPDISPPIVIPDDSKDCLMNQLIDPSDIDESSVMQLAEMGFPLEACRKAVYFTGNMGAEVAFNWIIVHMEEPDFAEPLTMPGYGGAASAGASVFGASGLDNQPPEEIVAIITSMGFQRNQAIQALRATNNNLERALDWIFSHPEFEEDSDFVIEMENNANANIISEAKPEGPRVKDGSGTYELFAFISHMGTSTMSGHYICHIKKEGRWVIYNDHKVCASERPPKDLGYMYFYRRIPS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.