Learn More
Description
Specifications
Specifications
| Product Type | cDNA |
| Gene | USP19 |
| Accession Number | NP_006668 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Gene ID (Entrez) | 10869 |
| Gene Alias | Deubiquitinating enzyme 19, EC 3.1.2.15, EC 3.4.19.12, KIAA0891, Ubiquitin Carboxyl-Terminal Hydrolase 19, ubiquitin specific peptidase 19, ubiquitin thioesterase 19, Ubiquitin thiolesterase 19, Ubiquitin-specific-processing protease 19, Zinc finger MYND domain-containing protein 9, ZMYND9, ZMYND9ubiquitin specific protease 19 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
