Learn More
Abnova™ Human USP21 Full-length ORF (AAH90946.1, 1 a.a. - 565 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
This gene encodes a member of the C19 peptidase family, also known as family 2 of ubiquitin carboxy-terminal hydrolases. The encoded protein cleaves ubiquitin from ubiquitinated proteins for recycling in intracellular protein degradation. The encoded protein is also able to release NEDD8, a ubiquitin-like protein, from NEDD8-conjugated proteins. This gene has been referred to as USP16 and USP23 but is now known as USP21. Alternatively spliced transcript variants have been described. [provided by RefSeq]
Specifications
Specifications
| Accession Number | AAH90946.1 |
| For Use With (Application) | Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 27005 |
| Molecular Weight (g/mol) | 89kDa |
| Name | Human USP21 Full-length ORF (AAH90946.1, 1 a.a. - 565 a.a.) Recombinant Protein with GST-tag at N-terminal |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 10 μg |
| Storage Requirements | Store at −80°C. Aliquot to avoid repeated freezing and thawing. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.