Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human USP21 Partial ORF (NP_036607, 466 a.a. - 565 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966793 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-793 25 μg
89-966-792 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-966-793 Supplier Abnova™ Supplier No. H00027005Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the C19 peptidase family, also known as family 2 of ubiquitin carboxy-terminal hydrolases. The encoded protein cleaves ubiquitin from ubiquitinated proteins for recycling in intracellular protein degradation. The encoded protein is also able to release NEDD8, a ubiquitin-like protein, from NEDD8-conjugated proteins. This gene has been referred to as USP16 and USP23 but is now known as USP21. Alternatively spliced transcript variants have been described. [provided by RefSeq]

Sequence: FSASRGSIKKSSVGVDFPLQRLSLGDFASDKAGSPVYQLYALCNHSGSVHYGHYTALCRCQTGWHVYNDSRVSPVSENQVASSEGYVLFYQLMQEPPRCL

Specifications

Accession Number NP_036607
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 27005
Molecular Weight (g/mol) 36.74kDa
Name USP21 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen FSASRGSIKKSSVGVDFPLQRLSLGDFASDKAGSPVYQLYALCNHSGSVHYGHYTALCRCQTGWHVYNDSRVSPVSENQVASSEGYVLFYQLMQEPPRCL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC3394/USP16/USP23
Common Name USP21
Gene Symbol USP21
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.