Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UTRN Partial ORF (NP_009055, 3328 a.a. - 3433 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966893 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-893 10 μg
89-966-894 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-966-893 Supplier Abnova™ Supplier No. H00007402Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene shares both structural and functional similarities with the dystrophin gene. It contains an actin-binding N-terminus, a triple coiled-coil repeat central region, and a C-terminus that consists of protein-protein interaction motifs which interact with dystroglycan protein components. The protein encoded by this gene is located at the neuromuscular synapse and myotendinous junctions, where it participates in post-synaptic membrane maintenance and acetylcholine receptor clustering. Mouse studies suggest that this gene may serve as a functional substitute for the dystrophin gene and therefore, may serve as a potential therapeutic alternative to muscular dystrophy which is caused by mutations in the dystrophin gene. Alternative splicing of the utrophin gene has been described; however, the full-length nature of these variants has not yet been determined. [provided by RefSeq]

Sequence: LEARMQILEDHNKQLESQLHRLRQLLEQPESDSRINGVSPWASPQHSALSYSLDPDASGPQFHQAAGEDLLAPPHDTSTDLTEVMEQIHSTFPSCCPNVPSRPQAM

Specifications

Accession Number NP_009055
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7402
Molecular Weight (g/mol) 37.4kDa
Name UTRN (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen LEARMQILEDHNKQLESQLHRLRQLLEQPESDSRINGVSPWASPQHSALSYSLDPDASGPQFHQAAGEDLLAPPHDTSTDLTEVMEQIHSTFPSCCPNVPSRPQAM
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DMDL/DRP/DRP1/FLJ23678
Common Name UTRN
Gene Symbol UTRN
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.