Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UVRAG Partial ORF (NP_003360.2, 601 a.a. - 699 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966900 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-900 10 μg
89-966-901 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-966-900 Supplier Abnova™ Supplier No. H00007405Q01S

Used for AP, Array, ELISA, WB-Re

This gene complements the ultraviolet sensitivity of xeroderma pigmentosum group C cells and encodes a protein with a C2 domain. The protein activates the Beclin1-PI(3)KC3 complex, promoting autophagy and suppressing the proliferation and tumorigenicity of human colon cancer cells. Chromosomal aberrations involving this gene are associated with left-right axis malformation and mutations in this gene have been associated with colon cancer. [provided by RefSeq]

Sequence: LPSEQAGSASVQLPGEFHPVSEAELCCTVEQAEEIIGLEATGFASGDQLEAFNCIPVDSAVAVECDEQVLGEFEEFSRRIYALNENVSSFRRPRRSSDK

Specifications

Accession Number NP_003360.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7405
Molecular Weight (g/mol) 36.63kDa
Name UVRAG (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen LPSEQAGSASVQLPGEFHPVSEAELCCTVEQAEEIIGLEATGFASGDQLEAFNCIPVDSAVAVECDEQVLGEFEEFSRRIYALNENVSSFRRPRRSSDK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DHTX/p63
Common Name UVRAG
Gene Symbol UVRAG
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.