Learn More
Abnova™ Human UXT Full-length ORF (NP_705582.1, 1 a.a. - 169 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
This gene encodes a novel protein which is highly conserved in mouse. It interacts with the N-terminus of the androgen receptor and plays a role in facilitating receptor-induced transcriptional activation. It is also likely to be involved in tumorigenesis as it is abundantly expressed in tumor tissues. This gene is part of a gene cluster on chromosome Xp11.23. Alternative splicing results in 2 transcript variants encoding different isoforms. [provided by RefSeq]
Specifications
Specifications
| Accession Number | NP_705582.1 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 8409 |
| Molecular Weight (g/mol) | 46kDa |
| Name | UXT (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 25 μg |
| Immunogen | MVFPLPTPQEPIMATPPKRRAVEATGEKVLRYETFISDVLQRDLRKVLDHRDKVYEQLAKYLQLRNVIERLQEAKHSELYMQVDLGCNFFVDTVVPDTSRIYVALGYGFFLELTLAEALKFIDRKSSLLTELSNSLTKDSMNIKAHIHMLLEGLRELQGLQNFPEKPHH |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.