Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UXT Partial ORF (NP_705582, 10 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966907 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-907 25 μg
89-966-906 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-966-907 Supplier Abnova™ Supplier No. H00008409Q01L

Used for AP, Array, ELISA, WB-Re

This gene encodes a novel protein which is highly conserved in mouse. It interacts with the N-terminus of the androgen receptor and plays a role in facilitating receptor-induced transcriptional activation. It is also likely to be involved in tumorigenesis as it is abundantly expressed in tumor tissues. This gene is part of a gene cluster on chromosome Xp11.23. Alternative splicing results in 2 transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: EPIMATPPKRRAVEATGEKVLRYETFISDVLQRDLRKVLDHRDKVYEQLAKYLQLRNVIERLQEAKHSELYMQVDLGCNFFVDTVVPDTSR

Specifications

Accession Number NP_705582
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8409
Molecular Weight (g/mol) 35.75kDa
Name UXT (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen EPIMATPPKRRAVEATGEKVLRYETFISDVLQRDLRKVLDHRDKVYEQLAKYLQLRNVIERLQEAKHSELYMQVDLGCNFFVDTVVPDTSR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ART-27
Common Name UXT
Gene Symbol UXT
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.