Learn More
Description
- Also known as: HPAO; SSAO; VAP1; VAP-1
- VAP1 is a member of the semicarbazide-sensitive amine oxidase family.
- It is a copper amine oxidase that catalyzes the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor.
- It is localized to the cell surface, has adhesive properties as well as monoamine oxidase activity, and may be involved in leukocyte trafficking.
- Alterations in levels of VAP1 may be associated with many diseases, including diabetes mellitus.
- Alternatively spliced transcripts encoding different proteins have been described.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hAOC3 |
| Accession Number | NP_003725 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2305 bp |
| Gene ID (Entrez) | 8639 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
