Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human VAP-1 Quantikine ELISA Kit
SDP

Catalog No. DVAP10 Shop All Novus Biologicals Products
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
DVAP10 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DVAP10 Supplier R&D Systems™ Supplier No. DVAP10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human VAP-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human VAP-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human VAP-1/AOC3 in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).

Specifications

Assay Range 0.78 - 50ng/mL
Assay Sensitivity 0.077ng/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Natural and Recombinant Human VAP-1
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 8639
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym amine oxidase, copper containing 3 (vascular adhesion protein 1), AOC3, Copper amine oxidase, EC 1.4.3, HPAO, HPAOSSAO, Semicarbazide-sensitive amine oxidase, SSAO, VAP1, VAP1EC 1.4.3.21, VAP-1 membrane primary amine oxidase, Vascular adhesion protein 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.