Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VARS Full-length ORF (AAH12808.1, 1 a.a. - 1264 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966936 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-936 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-966-936 Supplier Abnova™ Supplier No. H00007407P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. The protein encoded by this gene belongs to class-I aminoacyl-tRNA synthetase family and is located in the class III region of the major histocompatibility complex. [provided by RefSeq]

Sequence: MSTLYVSPHPDAFPSLRALIAARYGEAGEGPGWGGAHPRICLQPPPTSRTSFPPPRLPALEQGPGGLWVWGATAVAQLLWPAGLGGPGGSRAAVLVQQWVSYADTELIPAACGATLPALGLRSSAQDPQAVLGALGRALSPLEEWLRLHTYLAGEAPTLADLAAVTALLLPFRYVLDPPARRIWNNVTRWFVTCVRQPEFRAVLGEVVLYSGARPLSHQPGPEAPALPKTAAQLKKEAKKREKLEKFQQKQKIQQQQPPPGEKKPKPEKREKRDPGVITYDLPTPPGEKKDVSGPMPDSYSPRYVEAAWYPWWEQQGFFKPEYGRPNVSAANPRGVFMMCIPPPNVTGSLHLGHALTNAIQDSLTRWHRMRGETTLWNPGCDHAGIATQVVVEKKLWREQGLSRHQLGREAFLQEVWKWKEEKGDRIYHQLKKLGSSLDWDRACFTMDPKLSAAVTEAFVRLHEEGIIYRSTRLVNWSCTLNSAISDIEVDKKELTGRTLLSVPGYKEKVEFGVLVSFAYKVQGSDSDEEVVVATTRIETMLGDVAVAVHPKDTRYQHLKGKNVIHPFLSRSLPIVFDEFVDMDFGTGAVKITPAHDQNDYEVGQRHGLEAISIMDSRGALINVPPPFLGLPRFEARKAVLVALKERGLFRGIEDNPMVVPLCNRSKDVVEPLLRPQWYVRCGEMAQAASAAVTRGDLRILPEAHQRTWHAWMDNIREWCISRQLWWGHRIPAYFVTVSDPAVPPGEDPDGRYWVSGRNEAEAREKAAKEFGVSPDKISLQQDEDVLDTWFSSGLFPLSILGWPNQSEDLSVFYPGTLLETGHDILFFWVARMVMLGLKLTGRLPFREVYLHAIVRDAHGRKMSKSLGNVIDPLDVIYGISLQGLHNQLLNSNLDPSEVEKAKEGQKADFPAGIPECGTDALRFGLCAYMSQGRDINLDVNRILGYRHFCNKLWNATKFALRGLGKGFVPSPTSQPGGHESLVDRWIRSRLTEAVRLSNQGFQAYDFPAVTTAQYSFWLYELCDVYLECLKPVLNGVDQVAAECARQTLYTCLDVGLRLLSPFMPFVTEELFQRLPRRMPQAPPSLCVTPYPEPSECSWKDPEAEAALELALSITRAVRSLRADYNLTRIRPDCFLEVADEATGALASAVSGYVQALASAGVVAVLALGAPAPQGCAVALASDRCSIHLQLQGLVDPARELGKLQAKRVEAQRQAQRLRERRAASGYPVKVPLEVQEADEAKLQQTEAELRKVDEAIALFQKML

Specifications

Accession Number AAH12808.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7407
Molecular Weight (g/mol) 166.9kDa
Name VARS (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MSTLYVSPHPDAFPSLRALIAARYGEAGEGPGWGGAHPRICLQPPPTSRTSFPPPRLPALEQGPGGLWVWGATAVAQLLWPAGLGGPGGSRAAVLVQQWVSYADTELIPAACGATLPALGLRSSAQDPQAVLGALGRALSPLEEWLRLHTYLAGEAPTLADLAAVTALLLPFRYVLDPPARRIWNNVTRWFVTCVRQPEFRAVLGEVVLYSGARPLSHQPGPEAPALPKTAAQLKKEAKKREKLEKFQQKQKIQQQQPPPGEKKPKPEKREKRDPGVITYDLPTPPGEKKDVSGPMPDSYSPRYVEAAWYPWWEQQGFFKPEYGRPNVSAANPRGVFMMCIPPPNVTGSLHLGHALTNAIQDSLTRWHRMRGETTLWNPGCDHAGIATQVVVEKKLWREQGLSRHQLGREAFLQEVWKWKEEKGDRIYHQLKKLGSSLDWDRACFTMDPKLSAAVTEAFVRLHEEGIIYRSTRLVNWSCTLNSAISDIEVDKKELTGRTLLSVPGYKEKVEFGVLVSFAYKVQGSDSDEEVVVATTRIETMLGDVAVAVHPKDTRYQHLKGKNVIHPFLSRSLPIVFDEFVDMDFGTGAVKITPAHDQNDYEVGQRHGLEAISIMDSRGALINVPPPFLGLPRFEARKAVLVALKERGLFRGIEDNPMVVPLCNRSKDVVEPLLRPQWYVRCGEMAQAASAAVTRGDLRILPEAHQRTWHAWMDNIREWCISRQLWWGHRIPAYFVTVSDPAVPPGEDPDGRYWVSGRNEAEAREKAAKEFGVSPDKISLQQDEDVLDTWFSSGLFPLSILGWPNQSEDLSVFYPGTLLETGHDILFFWVARMVMLGLKLTGRLPFREVYLHAIVRDAHGRKMSKSLGNVIDPLDVIYGISLQGLHNQLLNSNLDPSEVEKAKEGQKADFPAGIPECGTDALRFGLCAYMSQGRDINLDVNRILGYRHFCNKLWNATKFALRGLGKGFVPSPTSQPGGHESLVDRWIRSRLTEAVRLSNQGFQAYDFPAVTTAQYSFWLYELCDVYLECLKPVLNGVDQVAAECARQTLYTCLDVGLRLLSPFMPFVTEELFQRLPRRMPQAPPSLCVTPYPEPSECSWKDPEAEAALELALSITRAVRSLRADYNLTRIRPDCFLEVADEATGALASAVSGYVQALASAGVVAVLALGAPAPQGCAVALASDRCSIHLQLQGLVDPARELGKLQAKRVEAQRQAQRLRERRAASGYPVKVPLEVQEADEAKLQQTEAELRKVDEAIALFQKML
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias G7A/VARS1/VARS2
Common Name VARS
Gene Symbol VARS
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.