Learn More
American Research Products Inc Human Vascular cell adhesion m
Supplier:  American Research Products Inc CSBE04753H
Encompass
Human Vascular cell adhesion molecule 1 VCAM-1 ELISA kit from CUSABIO Detection Range 1 563 ng/mL-100 ng/mL Sensitivity 0 611 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29130
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.