Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human VASH1 Synthetic Peptide

Catalog No. PEP1262
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1262 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1262 Supplier Invitrogen™ Supplier No. PEP1262
Add to Cart
Add to Cart

Synthetic Peptide

VASH1 Synthetic Peptide, PEP-1262, from Invitrogen.

VASH1 is a protein that possesses actin binding activity and metallocarboxypeptidase activity. It is involved in several processes including negative regulation of angiogenesis, negative regulation of blood vessel endothelial cell migration, and proteolysis. VASH1 acts upstream of or within processes such as negative regulation of endothelial cell migration, negative regulation of endothelial cell proliferation, and negative regulation of lymphangiogenesis. It is located in the apical part of the cell, endoplasmic reticulum, and extracellular space. VASH1 is implicated in liver cirrhosis and portal hypertension, and is a biomarker of liver cirrhosis. Diseases associated with VASH1 include Portal Hypertension and Myasthenic Syndrome, Congenital, 14.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 22846
Species Human
Purification Method purified
Accession Number Q7L8A9
Common Name Human VASH1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol VASH1
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.