Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VASP Full-length ORF (AAH38224.1, 1 a.a. - 380 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966946 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-946 25 μg
89-966-945 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-966-946 Supplier Abnova™ Supplier No. H00007408P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Vasodilator-stimulated phosphoprotein (VASP) is a member of the Ena-VASP protein family. Ena-VASP family members contain an EHV1 N-terminal domain that binds proteins containing E/DFPPPPXD/E motifs and targets Ena-VASP proteins to focal adhesions. In the mid-region of the protein, family members have a proline-rich domain that binds SH3 and WW domain-containing proteins. Their C-terminal EVH2 domain mediates tetramerization and binds both G and F actin. VASP is associated with filamentous actin formation and likely plays a widespread role in cell adhesion and motility. VASP may also be involved in the intracellular signaling pathways that regulate integrin-extracellular matrix interactions. VASP is regulated by the cyclic nucleotide-dependent kinases PKA and PKG. [provided by RefSeq]

Sequence: MSETVICSSRATVMLYDDGNKRWLPAGTGPQAFSRVQIYHNPTANSFRVVGRKMQPDQQVVINCAIVRGVKYNQATPNFHQWRDARQVWGLNFGSKEDAAQFAAGMASALEALEGGGPPPPPALPTWSVPNGPSPEEVEQQKRQQPGPSEHIERRVSNAGGPPAPPAGGPPPPPGPPPPPGPPPPPGLPPSGVPAAAHGAGGGPPPAPPLPAAQGPGGGGAGAPGLAAAIAGAKLRKVSKQEEASGGPTAPKAESGRSGGGGLMEEMNAMLARRRKATQVGEKTPKDESANQEEPEARVPAQSESVRRPWEKNSTTLPRMKSSSSVTTSETQPCTPSSSDYSDLQRVKQELLEEVKKELQKVKEEIIEAFVQELRKRGSP

Specifications

Accession Number AAH38224.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7408
Molecular Weight (g/mol) 66.2kDa
Name VASP (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MSETVICSSRATVMLYDDGNKRWLPAGTGPQAFSRVQIYHNPTANSFRVVGRKMQPDQQVVINCAIVRGVKYNQATPNFHQWRDARQVWGLNFGSKEDAAQFAAGMASALEALEGGGPPPPPALPTWSVPNGPSPEEVEQQKRQQPGPSEHIERRVSNAGGPPAPPAGGPPPPPGPPPPPGPPPPPGLPPSGVPAAAHGAGGGPPPAPPLPAAQGPGGGGAGAPGLAAAIAGAKLRKVSKQEEASGGPTAPKAESGRSGGGGLMEEMNAMLARRRKATQVGEKTPKDESANQEEPEARVPAQSESVRRPWEKNSTTLPRMKSSSSVTTSETQPCTPSSSDYSDLQRVKQELLEEVKKELQKVKEEIIEAFVQELRKRGSP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name VASP
Gene Symbol VASP
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.