Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VAT1 (NP_006364, 1 a.a. - 393 a.a.) Full-length Recombinant Protein with His-tag

Catalog No. 89966949 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-949 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-966-949 Supplier Abnova™ Supplier No. P4539
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

Synaptic vesicles are responsible for regulating the storage and release of neurotransmitters in the nerve terminal. The protein encoded by this gene is an abundant integral membrane protein of cholinergic synaptic vesicles and is thought to be involved in vesicular transport. It belongs to the quinone oxidoreductase subfamily of zinc-containing alcohol dehydrogenase proteins. [provided by RefSeq]

Sequence: MGSSHHHHHHSSGLVPRGSHMSDEREVAEAATGEDASSPPPKTEAASDPQHPAASEGAAAAAASPPLLRCLVLTGFGGYDKVKLQSRPAAPPAPGPGQLTLRLRACGLNFADLMARQGLYDRLPPLPVTPGMEGAGVVIAVGEGVSDRKAGDRVMVLNRSGMWQEEVTVPSVQTFLIPEAMTFEEAAALLVNYITAYMVLFDFGNLQPGHSVLVHMAAGGVGMAAVQLCRTVENVTVFGTASASKHEALKENGVTHPIDYHTTDYVDEIKKISPKGVDIVMDPLGGSDTAKGYNLLKPMGKVVTYGMANLLTGPKRNLMALARTWWNQFSVTALQLLQANRAVCGFHLGYLDGEVELVSGVVARLLALYNQGHIKPHIDSVWPFEKVADAMKQMQEKKNVGKVLLVPGPEKEN

Specifications

Accession Number NP_006364
Concentration 0.5mg/mL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 10493
Molecular Weight (g/mol) 44.1kDa
Name VAT1 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Quality Control Testing 15% SDS-PAGE Stained with Coomassie Blue
Quantity 50 μg
Immunogen MGSSHHHHHHSSGLVPRGSHMSDEREVAEAATGEDASSPPPKTEAASDPQHPAASEGAAAAAASPPLLRCLVLTGFGGYDKVKLQSRPAAPPAPGPGQLTLRLRACGLNFADLMARQGLYDRLPPLPVTPGMEGAGVVIAVGEGVSDRKAGDRVMVLNRSGMWQEEVTVPSVQTFLIPEAMTFEEAAALLVNYITAYMVLFDFGNLQPGHSVLVHMAAGGVGMAAVQLCRTVENVTVFGTASASKHEALKENGVTHPIDYHTTDYVDEIKKISPKGVDIVMDPLGGSDTAKGYNLLKPMGKVVTYGMANLLTGPKRNLMALARTWWNQFSVTALQLLQANRAVCGFHLGYLDGEVELVSGVVARLLALYNQGHIKPHIDSVWPFEKVADAMKQMQEKKNVGKVLLVPGPEKEN
Storage Requirements Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ20230/VATI
Common Name VAT1
Gene Symbol VAT1
Species E. coli
Recombinant Recombinant
Protein Tag His
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >90% by SDS - PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.