Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human VCAM-1/CD106 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP191177 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP191177 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP191177 Supplier Novus Biologicals™ Supplier No. NBP191177
Only null left
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human VCAM-1/CD106 ELISA Kit (Colorimetric) measures human VCAM-1/CD106 in cell culture supernatant, serum, plasma (edta, citrate, heparin).

Specifications

Assay Range 3.15 - 100ng/mL
Assay Sensitivity 0.6ng/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 7412
Gene Symbol VCAM1
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sVCAM-1, Conjugate Mixture containing Biotin-Conjugate anti-human sVCAM-1 monoclonal antibody mixed with Streptavidin-HRP, Human sVCAM-1 Standard lyophilized, 200ng/mL upon reconstitution, Control high, lyophilized, Control low, lyophilized, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1
Target VCAM-1/CD106
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.