Learn More
R&D Systems™ Human VCAM-1/CD106 Quantikine ELISA Kit
Description
Specifications
Specifications
| Assay Range | 6.25 - 200ng/mL |
| Assay Sensitivity | 1.26ng/mL |
| Conjugate | HRP |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
| Specificity | Natural and Recombinant Human sVCAM-1 |
| Sufficient For | 6 x 96-well Plates |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.