Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human VCAM-1 Recombinant Protein, PeproTech®

Catalog No. 15004250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15004250UG 250 μg
15004100UG 100 μg
150-04-10UG 10 μg
150-04-1MG 1 mg
15004500UG 500 μg
150-04-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 15004250UG Supplier Gibco™ Supplier No. 15004250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human VCAM-1 is a 74.1 kDa glycoprotein comprising the extracellular domain (674 amino acid residues) of VCAM-1. Monomeric glycosylated VCAM-1 migrates at an apparent molecular weight of approximately 90.0 kDa by SDS-PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P19320
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 7412
Molecular Weight (g/mol) 74.1 kDa
Name Human VCAM-1
Quantity 250 μg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B
Common Name VCAM-1 (CD106)
Gene Symbol VCAM1
Biological Activity Determined by its ability to support the adhesion of human U937 cells. The expected ED50 for this effect is 0.8-1.0 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence FKIETTPESR YLAQIGDSVS LTCSTTGCES PFFSWRTQID SPLNGKVTNE GTTSTLTMNP VSFGNEHSYL CTATCESRKL EKGIQVEIYSF PKDPEIHLSG PLEAGKPITV KCSVADVYPF DRLEIDLLKG DHLMKSQEFL EDADRKSLET KSLEVTFTPV IEDIGKVLVC RAKLHIDEMD SVPTVRQAVK ELQVYISPKN TVISVNPSTK LQEGGSVTMT CSSEGLPAPE IFWSKKLDNG NLQHLSGNAT LTLIAMRMED SGIYVCEGVN LIGKNRKEVE LIVQEKPFTV EISPGPRIAA QIGDSVMLTC SVMGCESPSF SWRTQIDSPL SGKVRSEGTN STLTLSPVS FENEHSYLCT VTCGHKKLEK GIQVELYSFPR DPEIEMSGGLV NGSSVTVSCK VPSVYPLDRLE IELLKGETILE NIEFLEDTDM KSLENKSLEMT FIPTIEDTGKA LVCQAKLHID DMEFEPKQRQ STQTLYVNVA PRDTTVLVSP SSILEEGSSV NMTCLSQGFP APKILWSRQL PNGELQPLSE NATLTLISTK MEDSGVYLCE GINQAGRSRK EVELIIQVTP KDIKLTAFPS ESVKEGDTVI ISCTCGNVPE TWIILKKKAE TGDTVLKSID GAYTIRKAQL KDAGVYECES KNKVGSQLRS LTLDVQGREN NKDYFSP
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 97% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.