Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human VEGF-B Recombinant Protein, PeproTech®

Catalog No. 10020B100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10020B100UG 100 μg
100-20B-1MG 1 mg
10020B20UG 20 μg
10020B250UG 250 μg
10020B500UG 500 μg
100-20B-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 10020B100UG Supplier Gibco™ Supplier No. 10020B100UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human VEGF-B is a 38.0 kDa, disulfide-linked homodimeric protein consisting of two 167 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Vascular endothelial growth factors (VEGFs) are a family of closely related growth factors having a conserved pattern of eight cysteine residues and sharing common VEGF receptors. VEGFs stimulate endothelial cells, induce angiogenesis, promote cell migration, increase vascular permeability, and inhibit apoptosis. VEGFB has structural similarities to VEGF and PIGF and is frequently co-expressed with VEGF. There are two alternatively spliced isoforms of VEGFB: VEGFB 167 and VEGFB 186. VEGFB 167, a highly basic heparin-binding protein, remains with the cell or extracellular matrix whereas, VEGFB 186 is readily secreted. VEGFB stimulates endothelial cell proliferation. VEGFB binds to the tyrosine kinase receptor VEGFR1 (flt1) and does not bind to VEGFR2. Vascular Endothelial Growth Factor B is widely expressed but is most abundant in heart, skeletal muscle, and pancreas. It has been suggested that VEGFB expression in human primary breast cancers is associated with lymph node metastasis. The genes that encode VEGFB have been mapped to chromosome 11q13.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49765
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 7423
Molecular Weight (g/mol) 38 kDa
Name Human VEGF-B
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias flt-1/VEGFR1 ligand; vascular endothelial growth factor B; vascular endothelial growth factor B167 precursor; vascular endothelial growth factor B186 precursor; vascular endothelial growth factor-B; Vegfb; VEGF-B; VEGFL; VEGF-related factor; Vrf
Common Name VEGFB
Gene Symbol VEGFB
Biological Activity Determined by the dose-dependent stimulation of the proliferation of human umbilical vein endothelial cells (HUVEC) in the presence of human VEGF 165. The expected ED50 for this effect is 1.0-2.0 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence PVSQPDAPGH QRKVVSWIDV YTRATCQPRE VVVPLTVELM GTVAKQLVPS CVTVQRCGGC CPDDGLECVP TGQHQVRMQI LMIRYPSSQL GEMSLEEHSQ CECRPKKKDS AVKPDSPRPL CPRCTQHHQR PDPRTCRCRC RRRSFLRCQG RGLELNPDTC RCRKLRR
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.