Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human VEGF-C Quantikine ELISA Kit
SDP

Catalog No. DVEC00 Shop All Novus Biologicals Products
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
DVEC00 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DVEC00 Supplier R&D Systems™ Supplier No. DVEC00
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human VEGF-C Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human VEGF-C Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human VEGF-C in cell culture supernates (50 ul), serum (10 ul), edta plasma (50 ul), platelet-poor plasma (50 ul), heparin plasma (50 ul), saliva (50 ul).

Specifications

Assay Range 109.00 - 7000pg/mL
Assay Sensitivity 48.4pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Platelet-poor Plasma, Saliva, Serum
Specificity Natural Human VEGF-C, Recombinant Human VEGF-C (wild type), and Recombinant Human VEGF-C (Cys156Ser substituted)
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 7424
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (50uL),Platelet-poor Plasma (50uL),Heparin Plasma (50uL),Saliva (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGFC, VEGF-C, VRPFLT4 ligand DHM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.