Learn More
Gibco™ Human VEGF-C Recombinant Protein, PeproTech®
Description
Recombinant Human VEGF-C is a disulfide-linked homodimeric protein consisting of two 13.5 kDa polypeptide chains of 116 amino acid residues. Due to glycosylation, the protein migrates as a 30.0-33.0 kDa band by SDS-PAGE analysis under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P49767 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 7424 |
| Molecular Weight (g/mol) | 26.2 kDa |
| Name | Human VEGF-C |
| Quantity | 250 μg |
| Source | HEK293 cells |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.