Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human VEGF-D Recombinant Protein, PeproTech®

Catalog No. 10020D250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10020D250UG 250 μg
10020D100UG 100 μg
10020D10UG 10 μg
100-20D-1MG 1 mg
100-20D-2UG 2 μg
10020D500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 10020D250UG Supplier Gibco™ Supplier No. 10020D250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human VEGF-D is a 26.2 kDa, non-disulfide linked, homodimeric protein consisting of two 117 amino acid polypeptide chains. Due to glycosylation, the protein migrates as a 20.0-22.0 kDa band by SDS-PAGE analysis under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family and is active in angiogenesis, lymphangiogenesis, and endothelial cell growth. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-2 and VEGFR-3 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor C.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43915
For Use With (Application) Control, ELISA Standard, Binding Assay, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 2277
Molecular Weight (g/mol) 26.2 kDa
Name Human VEGF-D
Purification Method Purified
Quantity 250 μg
Source HEK293 cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias AI325264; c-fos induced growth factor; c-fos induced growth factor (vascular endothelial growth factor D); c-fos-induced growth factor; FIGF; Vascular endothelial growth factor D; vascular endothelial growth factor-D; VEGF4; VEGFD; VEGF-D; VEGFR2 and -3 ligand
Common Name VEGFD
Gene Symbol VEGFD
Biological Activity Measured by its ability to bind recombinant human VEGFR3/Flt-4 Fc in a functional ELISA.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence FAATFYDIET LKVIDEEWQR TQCSPRETCV EVASELGKST NTFFKPPCVN VFRCGGCCNE ESLICMNTST SYISKQLFEI SVPLTSVPEL VPVKVANHTG CKCLPTAPRH PYSIIRR
Content And Storage -20°C
Activity Measured by its ability to bind recombinant human VEGFR3/Flt-4 Fc in a functional ELISA.
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.