Learn More
Abcam Human VEGF ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2225101X96TESTS
Encompass_Preferred
Human VEGF ELISA kit is a single-wash, 90-minute SimpleStep ELISA used to quantify human vascular endothelial growth factor (VEGF) with a sensitivity of 2.7 pg/mL. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA with a 450 nm readout; compatible with any standard plate reader - Different formats for different needs including 384-well plate for higher throughput - Design your own immunoassay: we also offer the cojugation-ready antibody pair - Cited in over 30 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2824966